Imprimer
Tellurium standard solution, for ICP, 30% HCl, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PTEH-100 VHG-PTEH-500 VHG-TTEH-100 VHG-TTEH-500
PROOVHG-PTEH-100EA 193 EUR
PROOVHG-PTEH-100 PROOVHG-PTEH-500 PROOVHG-TTEH-100 PROOVHG-TTEH-500
Tellurium standard solution, for ICP, 30% HCl, CRM, VHG
Étalons Étalons cationiques Standards de tellure
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.