Imprimer
Volumetric flasks, class A, glass, amber, NS glass stopper, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2.37445 2.37447 2.37448 2.37450 2.37451 2.37452 2.37443 2.37453 2.37446 2.37438
BRND2.37445EA 86.4 EUR
BRND2.37445 BRND2.37447 BRND2.37448 BRND2.37450 BRND2.37451 BRND2.37452 BRND2.37443 BRND2.37453 BRND2.37446 BRND2.37438
Volumetric flasks, class A, glass, amber, NS glass stopper, BLAUBRAND®
Ballons Fioles jaugées
Borosilicate glass 3.3, amber, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
  • Total order from one batch, supplied with batch certificate
*Wide neck
Certifications: DIN EN ISO 1042