Imprimer
Titanium standard solution, for ICP, in HNO₃, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PTINF-100 VHG-LTINF-100 VHG-PTINF-50 VHG-PTINF-500 VHG-TTINF-100 VHG-TTINF-500
PROOVHG-PTINF-100EA 164 EUR
PROOVHG-PTINF-100 PROOVHG-LTINF-100 PROOVHG-PTINF-50 PROOVHG-PTINF-500 PROOVHG-TTINF-100 PROOVHG-TTINF-500
Titanium standard solution, for ICP, in HNO₃, CRM, VHG
Étalons Étalons cationiques Standards de titane
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.