Imprimer
Europium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PEUN-100 VHG-PEUN-500 VHG-TEUN-100 VHG-TEUN-500
PROOVHG-PEUN-100EA 209 EUR
PROOVHG-PEUN-100 PROOVHG-PEUN-500 PROOVHG-TEUN-100 PROOVHG-TEUN-500
Europium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Étalons Étalons cationiques Standards d'europium
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.