Imprimer
Erbium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Fournisseur: LGC Standards
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PROOVHG-PERN-100EA 183 EUR
PROOVHG-PERN-100 PROOVHG-TERN-100 PROOVHG-PERN-50 PROOVHG-PERN-500 PROOVHG-TERN-500
Erbium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Étalons Étalons cationiques Standards d'erbium
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.