Imprimer
Dysprosium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PDYN-100 VHG-PDYN-500 VHG-TDYN-100 VHG-TDYN-500
PROOVHG-PDYN-100EA 193 EUR
PROOVHG-PDYN-100 PROOVHG-PDYN-500 PROOVHG-TDYN-100 PROOVHG-TDYN-500
Dysprosium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Étalons Étalons cationiques Standards de dysprosium
These solutions are intended for use as a certified reference material (CRM) or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.