Imprimer
Push button safety covers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
104602 104603
BRDY104602EA 112 EUR
BRDY104602 BRDY104603
Push button safety covers
Cadenas de sécurité
These devices made from metal and clear PC guard against unauthorised operation or accidental activation.
  • Clear Lexan cover with hinged door can be kept open for easy access when not in use
  • For use with button style IEC 22,5 mm or NEMA 30,5 mm