Imprimer
Laboratory bottles, narrow neck, with high temperature closures, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
218012417 218013619 218014418 218015414 218011712 218016316 218017312 218018617
215-4127EA 23.1 EUR
215-4127 215-4128 215-4129 215-4130 SCOT218011712 215-4131 215-4132 215-4133
Laboratory bottles, narrow neck, with high temperature closures, DURAN®
Flacons Flacons en verre
DURAN® original laboratory bottles are manufactured of DURAN® borosilicate 3.3 glass, clear, renowned for its excellent thermal performance and high resistance to chemical attack.
  • Graduated in nature
  • Suitable for dry heat sterilisation
The presence of permanent retrace code on each bottle for full manufacturing lot traceability. The addition of 30% glass fibre reinforcement to the PBT resin increases stiffness, tensile strength and impact resistance of the screw cap.
Certifications: Conforms to USP/EP Type I and international food contact regulations such as (EU) No 10/2011.
Informations de livraison: Supplied with PBT cap with PTFE faced silicone liner and ETFE pouring rings.