Imprimer
Arsenic standard solutions, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-SPAS3-100 VHG-SPAS5W-100 VHG-SPAS3-50 VHG-SPAS5W-50
PROOVHG-SPAS3-100EA 167 EUR
PROOVHG-SPAS3-100 PROOVHG-SPAS5W-100 PROOVHG-SPAS3-50 PROOVHG-SPAS5W-50
Arsenic standard solutions, VHG
Étalons Étalons cationiques Standards d'arsenic
This standard solution is intended for use as a calibration or reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.
Certifications: Accredited to ISO 9001 and ISO/IEC 17025. Traceable to NIST and SRM.