Imprimer
Economy spill kit
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
813858 813857
121-0190EA 72.4 EUR
121-0190 121-0189
Economy spill kit
Matériaux absorbants
Economy spill kit for chemicals are designed for small spills of chemicals and hazardous liquids while kit for maintanance are designed for small spills of industrial liquids including oils, water, solvents, coolants. This kit is ideal for transport of small quantities of liquids.
  • Compact in size
  • Requires small storage area
  • Water-resistant
  • Lightweight bag keeps sorbents dry
Informations de livraison: Supplied with 10 pads (410×510 mm), 2 socs (Ø 76×1220 mm), one pair of gloves and one disposable bag.
Cdt: Packed in a yellow PVC bag with handles.