Imprimer
Buckets, with leaktight lid, EUROTUBO®
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
222802 222803 222804 222805
216-1651EA 9.4 EUR
216-1651 216-1652 216-1653 216-1654
Buckets, with leaktight lid, EUROTUBO®
Godets
These disposable buckets are made of polypropylene with tight fitting and leakproof lids.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
All buckets are supplied with a sturdy and convenient white plastic handle for easier carrying.