Imprimer
Elemental analysis standards, US EPA 23 metals standard, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-SM23-100 VHG-SM23-500
PROOVHG-SM23-100EA 1260 EUR
PROOVHG-SM23-100 PROOVHG-SM23-500
Elemental analysis standards, US EPA 23 metals standard, VHG
Étalons Étalons cationiques Standards multi-éléments
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as inductively coupled plasma optical emission spectroscopy (ICP-OES) and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi element aqeous standards for Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Tl, V, Zn at 100 µg/ml in 5% HNO₃, trace tartaric acid, trace HF.
Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.