Imprimer
High-density shelving system, Super Erecta™ Top-Track™
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TTE18K4 TTM18K4
METOTTE18K4EA 0 EUR
METOTTE18K4 METOTTM18K4
High-density shelving system, Super Erecta™ Top-Track™
Mobilier Étagères
Top-Track™ shelving system increases storage capacity up to 50% and maximises the usable storage area in a given space.
  • Maximum load rating for a single unit is 408,2 kg
  • Stainless steel units provide maximum protection in corrosive environments
The overhead track system guides the mobile units and opens an access aisle between any two units. The track is above the storage units enabling easy access.
Stationary end unit kits include four 2184 mm high posts and the necessary stainless-steel hardware for connecting track sets to two stationary end units in double deep configuration. The stationary end unit must contain a minimum of four shelves. The width of the stationary end units should be equal to or greater than the width of the mobile units.
Mobile unit kits include four 1880 mm posts, four chrome or stainless steel casters with bumpers, four caster channels, and four roller assemblies. For the mobile units to function properly, the floor area should be level, smooth, and free from large cracks and raised obstacles.
Informations de commande: Super Erecta™ shelves must be purchased separately. Additional accessories such as shelf dividers, stackable ledges, tray slides and label holders need to be ordered separately.