Imprimer
Security unit for shelving systems, Super Erecta®
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SEC33S SEC35S
METOSEC33SEA 5730 EUR
METOSEC33S METOSEC35S
Security unit for shelving systems, Super Erecta®
Mobilier Étagères
The stainless steel stem caster unit protect valuable materials and sensitive items from loss and they are ideal for use in wet environments. The heavy-gauge open wire construction keeps contents visible at all time making it easy to check inventory.
  • Shelves can be positioned in 25 mm increments along the entire height of post
  • Double doors open 270 degrees and can be secured along the sides of the unit
Informations de commande: The casters and intermediate shelves are not included with base models and must be ordered separately.