Imprimer
Calcium chlorure dihydraté USP, JP, Ph. Eur., BP testé pour les endotoxines
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Calcium chlorure à 2 molécules d'eau
6495.1000EA 77.1 EUR
6495.1000 6495.9025
Calcium chlorure dihydraté USP, JP, Ph. Eur., BP testé pour les endotoxines
Calcium chlorure dihydraté
Appearance: White or almost white, crystalline mass or colourless crystals.
Attention: Not for use as Active Pharmaceutical Ingredient (API)
Formule: CaCl₂·2H₂O
Poids moléculaire: 147.01 g/mol
Point d'ébullition: 1600
Point de fusion: 176 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00149613
Numéro CAS: 10035-04-8
EINECS: 233-140-8
Indice Merck: 13,01660
Commander maintenant
Résultats des tests de spécification
Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) [JP] ≤10 ppm
Bacterial endotoxins ≤2.5 EU/g
Residual solvents Passes test
As (Arsenic) [JP] ≤2 ppm
Cd (Cadmium) ≤0.1 ppm
Co (Cobalt) ≤1 ppm
Hg (Mercury) ≤2 ppm
Ni (Nickel) ≤6 ppm
Pb (Lead) ≤5 ppm
V (Vanadium) ≤10 ppm
Conforms to Ph.Eur./USP/BP/JP Passes test
Ph.Eur./BP specifications: .
Assay [Ph.Eur./BP] 97.0 - 103.0 %
Identification A Passes test USP
Identification B Passes test USP
Identification C Passes test Ph.Eur./BP
Appearance of solution Passes test Ph.Eur./BP
Acidity or alkalinity Passes test Ph.Eur./BP
SO4 (Sulphate) [Ph.Eur./BP] ≤300 ppm
Al (Aluminium) Passes test Ph.Eur./BP
Fe (Iron) [Ph.Eur./BP] ≤10 ppm
Magnesium and alkali metals [Ph.Eur./BP] ≤0.5 %
USP specifications: .
Assay [USP] 99.0 - 107.0 %
Iron, aluminium and phosphate Passes test JP
Limit of Mg and alkali salts [USP] ≤1.0 %
pH (25°C; 50 mg/ml) [USP] 4.5 - 9.2
JP specifications: .
Assay [JP] 96.7 - 103.3 %
Identification Passes test JP
pH (1 g/20 ml) [JP] 4.5 - 9.2
Clarity and colour of solution Passes test JP
SO4 (Sulphate) [JP] ≤0.024 %
Hypochlorite Passes test JP
Ba (Barium) Passes test JP