Imprimer
Nitrate(N), ion chromatography standard, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-INO3N-500 VHG-INO3N-100
PROOVHG-INO3N-500EA 360 EUR
PROOVHG-INO3N-500 PROOVHG-INO3N-100
Nitrate(N), ion chromatography standard, VHG
Étalons Étalons anioniques Standards de nitrate(N)
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and SRM.