Imprimer
Nitrate ion chromatography standard, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-I1PNO3-100 VHG-INO3-100 VHG-INO3-500
PROOVHG-I1PNO3-100EA 301 EUR
PROOVHG-I1PNO3-100 PROOVHG-INO3-100 PROOVHG-INO3-500
Nitrate ion chromatography standard, VHG
Étalons Étalons anioniques Standards de nitrate
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and SRM.