Imprimer
Cryoboxes, Plastic Coated, 148×148 mm
Fournisseur: RATIOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5144130 5144131
RATI5144130EA 69.7 EUR
RATI5144130 RATI5144131
Cryoboxes, Plastic Coated, 148×148 mm
Boîtes Boîtes pour cryotubes
Cryoboxes, cryo resistant cardboard.
  • Various combinations possible with different grid inserts
  • Height stated is the interior height
  • Supplied with lid
  • Cryo boxes can be used for larger vessels when grid is removed
  • Temperature resistant down to – 80 °C
For storage of cryovials at low temperatures.