Imprimer
Fluoride ion chromatography standards, CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-I1PF-500 VHG-IF1K-100 VHG-IF1K-500 VHG-IF100-500
PROOVHG-I1PF-500EA 872 EUR
PROOVHG-I1PF-500 PROOVHG-IF1K-100 PROOVHG-IF1K-500 PROOVHG-IF100-500
Fluoride ion chromatography standards, CRM, VHG
Étalons Étalons anioniques Standards de fluorure
This standard solution is intended for use as a certified reference material and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and SRM.