Imprimer
Human peripheral whole blood
Fournisseur: Charles River Laboratories Cell Solutions
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PB000F-1 PB000F-2
PB000F-1EA 0 EUR
PB000F-1 PB000F-2
Human peripheral whole blood
Sang
Whole blood is human blood from a standard blood donation. Whole blood is collected from IRB consented healthy human donors in FDA-registered collection centres. Whole blood is collected in a CPD collection bag.
Storage: Use immediately upon receipt.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.