Imprimer
Crystallising Dishes, with Spout
Fournisseur: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
75150 75151 75152 75153 75154 75155
TSLA75150EA 49.9 EUR
TSLA75150 TSLA75151 TSLA75152 TSLA75153 TSLA75154 TSLA75155
Crystallising Dishes, with Spout
Plateaux/Récipients/Pots Cristallisoirs
Temperature-resistant crystallising dishes made from transparent quartz glass.
  • Excellent chemical resistance
  • High resistance against thermal shock
  • Made for tasks up to 1050 °C
  • High purity
The TQS quartz glass crystallising dishes are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterise these products. Quartz glass products have a long life for frequent use with temperatures up to 1050 °C, but the durability is dependant upon use and applications.