Imprimer
Cell preservation media, HypoThermosol®
Marques : Charles River Laboratories Cell Solutions
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BPM101102 BPM101104 BPM101204
101102.EA 0 EUR
101102. 101104. 101204.
Cell preservation media, HypoThermosol®
Suppléments et additifs pour culture cellulaire
HypoThermosol® biopreservation media is a novel, engineered, optimised hypothermic storage and shipping media product. Serum-free, protein-free HypoThermosol® is designed to provide maximum storage and shipping stability for biologics at 2 to 8 °C.
This proprietary, optimised formulation mitigates temperature-induced molecular cell stress responses that occur during chilling and re-warming of biologics, intermediate products, and final cell products intended for research and clinical applications.
HypoThermosol® FRS includes components that scavenge free radicals, provide pH buffering, oncotic/osmotic support, energy substrates, and ionic concentrations that balance the intracellular state at low temperatures.
This reducing post-preservation necrosis and apoptosis as compared to commercial and home-brew isotonic and extracellular formulations. This results in greatly extended shelf life and improved post-preservation viability.
Certifications: Meets USP (71) Sterility and USP (85) Endotoxin testing standards, and is manufactured under cGMP.
Attention: Research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.