Imprimer
SkanIt™ Upgrade package, version 5.0
Fournisseur: THERMO LABSYSTEMS LIFE SCIENCE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N17646 N18600
LABLN17646EA 3440 EUR
LABLN17646 LABLN18600
SkanIt™ Upgrade package, version 5.0
Software Laboratory Software
This SkanIt Software upgrade package is specifically designed for Thermo Scientific Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers to enable the use of SkanIt software and USB connection between instrument and PC. The upgrade package is also compatible with the discontinued Fluoroskan Ascent, Fluoroskan Ascent FL, and Luminoskan Ascent microplate readers. SkanIt Software is available in two editions: a research edition for scientists working in life science research and a drug discovery edition with features for compliancy with FDA 21 CFR Part 11. Note: the upgrade package is compatible with Ascent instruments that are equipped with internal software version 2.6.
  • Compatible with all modern PC computers (Windows™ 7, 8.1, or 10; 64-bit version)
  • Converter module allows easy USB connection between instrument and PC
  • Helps prevent data loss due to modern data security systems
Powerful operation and data analysis
SkanIt Software offers a comprehensive selection of built-in calculations, including blank subtraction, standard curves, and concentration/dose-response curve fitting, classification, quality control, and kinetic and spectral analysis. SkanIt Software is an open-license platform that authorizes unlimited installation privileges. You can create measurement sessions and analyze results on your computer inside or outside of the lab. The software is available in nine languages: English, French, German, Spanish, Portuguese, Italian, Russian, Japanese, and Chinese.
Modern and intuitive user interface
A user-friendly session structure simplifies the measurement setup. The Virtual Pipetting Tool lets you quickly enter the plate sample information in a manner that resembles reagent pipetting in the laboratory, making it easy to visualize and define the plate layout. Data is easily exported from SkanIt Software: with a single mouse click, your data is available in Microsoft™ Excel™ for further analysis.
SkanIt cloud library
The built-in SkanIt cloud library provides an extensive collection of ready-made measurement sessions for various applications and measurement technologies. You can use these sessions as a template for your own assays or use them without modification for a quick start.
Informations de commande: Includes SkanIt Software version 5.0, SkanIt converter module, required cables, and upgrading instructions.