Imprimer
Chlorate ion chromatography standards, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-ICLO3-100 VHG-ICLO3-500
PROOVHG-ICLO3-100EA 190 EUR
PROOVHG-ICLO3-100 PROOVHG-ICLO3-500
Chlorate ion chromatography standards, VHG
Étalons Étalons anioniques Standards de chlorate
This solution is intended for use as a certified reference material and calibration standard, for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and CRM.