Imprimer
Rubidium standard solution, for ICP, in 5% HNO₃, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PRBN-100 VHG-PRBN-50 VHG-PRBN-500 VHG-TRBN-100 VHG-TRBN-500
PROOVHG-PRBN-100EA 190 EUR
PROOVHG-PRBN-100 PROOVHG-PRBN-50 PROOVHG-PRBN-500 PROOVHG-TRBN-100 PROOVHG-TRBN-500
Rubidium standard solution, for ICP, in 5% HNO₃, VHG
Étalons Étalons cationiques Standards de rubidium
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.