Imprimer
Germanium standard solution, for ICP, in 5% HNO₃, tr. HF, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PGENF-100 VHG-PGENF-500 VHG-TGENF-100 VHG-TGENF-500
PROOVHG-PGENF-100EA 193 EUR
PROOVHG-PGENF-100 PROOVHG-PGENF-500 PROOVHG-TGENF-100 PROOVHG-TGENF-500
Germanium standard solution, for ICP, in 5% HNO₃, tr. HF, VHG
Étalons Étalons cationiques Standards de germanium
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.