Imprimer
Gadolinium standard solution, for ICP, in 5% HNO₃, VHG
Fournisseur: LGC Standards
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PROOVHG-TGDN-100EA 337 EUR
PROOVHG-TGDN-100 PROOVHG-TGDN-500 PROOVHG-PGDN-500 PROOVHG-PGDN-50 PROOVHG-PGDN-100
Gadolinium standard solution, for ICP, in 5% HNO₃, VHG
Étalons Étalons cationiques Standards de gadolinium
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.