Imprimer
Iron standard solution, for ICP, in 5% HNO₃, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PFEN-100 VHG-PFEN-500 VHG-TFEN-100 VHG-PFEN-50 VHG-TFEN-500
PROOVHG-PFEN-100EA 164 EUR
PROOVHG-PFEN-100 PROOVHG-PFEN-500 PROOVHG-TFEN-100 PROOVHG-PFEN-50 PROOVHG-TFEN-500
Iron standard solution, for ICP, in 5% HNO₃, VHG
Étalons Étalons cationiques Standards de fer
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.