Imprimer
Aluminium standard solution, for ICP, in 5% HCl, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PALH-100 VHG-TALH-100 VHG-TALH-500 VHG-PALH-500
PROOVHG-PALH-100EA 170 EUR
PROOVHG-PALH-100 PROOVHG-TALH-100 PROOVHG-TALH-500 PROOVHG-PALH-500
Aluminium standard solution, for ICP, in 5% HCl, VHG
Étalons Étalons cationiques Standards d'aluminium
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.