Imprimer
Chromium standard solution, for ICP, in 5% HCl, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PCRH-100 VHG-PCRH-500 VHG-TCRH-100 VHG-TCRH-500
PROOVHG-PCRH-100EA 170 EUR
PROOVHG-PCRH-100 PROOVHG-PCRH-500 PROOVHG-TCRH-100 PROOVHG-TCRH-500
Chromium standard solution, for ICP, in 5% HCl, VHG
Étalons Étalons cationiques Standards de chrome
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.