Imprimer
Bromide ion chromatography standards, bromide standard, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-IBR-100 VHG-IBR-500
PROOVHG-IBR-100EA 190 EUR
PROOVHG-IBR-100 PROOVHG-IBR-500
Bromide ion chromatography standards, bromide standard, VHG
Étalons Étalons anioniques Standards de bromure
These standard solutions are intended for use as a calibration or reference standard for ion chromatography (IC) or techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and CRM.