Imprimer
Acetate standard, ion chromatography CRM, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-IACET-100 VHG-IACET-500
PROOVHG-IACET-100EA 190 EUR
PROOVHG-IACET-100 PROOVHG-IACET-500
Acetate standard, ion chromatography CRM, VHG
Étalons Étalons anioniques Standards d'acétate
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and CRM.