Imprimer
Iridium standard solution, for ICP, in 20% HCl, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VHG-PIRH-100 VHG-PIRH-500 VHG-TIRH-100 VHG-TIRH-500
PROOVHG-PIRH-100EA 369 EUR
PROOVHG-PIRH-100 PROOVHG-PIRH-500 PROOVHG-TIRH-100 PROOVHG-TIRH-500
Iridium standard solution, for ICP, in 20% HCl, VHG
Étalons Étalons cationiques Standards d'iridium
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certifications: Traceable to NIST, SRM.