Imprimer
Mouse recombinant IFN alpha 2 (from E. coli)
Fournisseur: ProSci
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PRSI92-577EA 306 EUR
PRSI92-577
Mouse recombinant IFN alpha 2 (from E. coli)
Protéines et Peptides
At least 23 different variants of Interferon- alpha are known. The individual proteins have molecular masses between 19-26 kD and consist of proteins with lengths of 156-166 and 172 amino acids. All IFN- alpha subtypes possess a common conserved sequence region between amino acid positions 115-151 while the amino-terminal ends are variable. Many IFN- alpha subtypes differ in their sequences at only one or two positions. Naturally occurring variants also include proteins truncated by 10 amino acids at the carboxyl-terminal end.
This recombinant protein can be used for biological assays. For research use only.