Imprimer

Anti-DLK1 Rat Polyclonal Antibody

Fournisseur: ProSci
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

PRSI36-291EA 454 EUR
PRSI36-291
Anti-DLK1 Rat Polyclonal Antibody
Anticorps
DLK1 (Protein delta homolog 1; Preadipocyte factor 1; Pref-1) is a regulator of adipocyte differentiation found in serum and urine. DLK1 might be a decoy ligand. Serum or plasma levels of soluble DLK1 affect negatively or positively adipogenesis and control mesenchymal cell fate. DLK1 is frequently upregulated in myelodysplastic syndrome (MDS) patients compared to non-leukemic individuals.

ELISA: (direct and indirect: 1:2,000-1:10,000). Flow Cytometry: (1:100-1:500). Western Blot: (1:2,000-1:10,000 using ECL. Suggested blocking and dilution buffer is TBST containing 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: DLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR