Imprimer
ESD safety shoes for ladies, lace-up, New Charcoal
Fournisseur: MILLE S.A.S
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NHBG3/45 NHBG3/39 NHBG3/40 NHBG3/42 NHBG3/44 NHBG3/46 NHBG3/47 NHBG3/43
MILENHBG3/45EA 107 EUR
MILENHBG3/45 MILENHBG3/39 MILENHBG3/40 MILENHBG3/42 MILENHBG3/44 MILENHBG3/46 MILENHBG3/47 MILENHBG3/43
ESD safety shoes for ladies, lace-up, New Charcoal
Chaussures et bottes
These anti-static, metal-free safety shoes are made from a very sturdy, water and oil resistant suede leather upper (2-2, 2 mm) plus an extra light, PU, dual-density outer sole.
  • Abrasion resistant, antibacterial Poromax lining giving high water-absorbing capacity for additional breathability
  • Waterproof tongue and heel grip
  • Textile, puncture resistant midsole (1430 N)
  • Toe protection cap (240 J), made from fibre glass, insulates from warmth and cold
Suitable for industry, finishing construction and logistics.
Colour: Charcoal Grey
Certifications: EN ISO 20345; IEC EN 61340-5-1 (ESD); S3, SRC.