Imprimer
Safety shoes, lace-up, Mycity
Fournisseur: MILLE S.A.S
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MCAA3/39 MCAA3/41 MCAA3/42 MCAA3/43 MCAA3/44 MCAA3/45 MCAA3/46 MCAA3/47 MCAN3/39 MCAA3/40 MCAN3/42 MCAN3/40 MCAN3/43 MCAN3/44 MCAN3/45 MCAN3/46 MCAN3/47 MCAN3/41
MILEMCAA3/39EA 134 EUR
MILEMCAA3/39 MILEMCAA3/41 MILEMCAA3/42 MILEMCAA3/43 MILEMCAA3/44 MILEMCAA3/45 MILEMCAA3/46 MILEMCAA3/47 MILEMCAN3/39 MILEMCAA3/40 MILEMCAN3/42 MILEMCAN3/40 MILEMCAN3/43 MILEMCAN3/44 MILEMCAN3/45 MILEMCAN3/46 MILEMCAN3/47 MILEMCAN3/41
Safety shoes, lace-up, Mycity
Chaussures et bottes
These metal-free safety shoes are made from full grain leather upper plus a very light, flexible, high performance PU sole with outstanding grip.
  • Mesh 3-D, breathable lining and heel grip
  • Outer sole made from innovative, anti-abrasion material
  • Textile, puncture resistant midsole
  • Toe protection cap (240 J) made from fibre glass insulates from warmth and cold
Suitable for industry, finishing construction and logistics.
Colour: Brown/black
Certifications: EN ISO 20345:2011; S3, SRC.