Imprimer
Refrigerated and heating circulators, CBC series
Fournisseur: IKA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4167000 4165000
IKAA4167000EA 9250 EUR
IKAA4167000 IKAA4165000
Refrigerated and heating circulators, CBC series
Thermostats à circulation Systèmes à circulation ouverte Cryothermostats à circulation
The CBC 5 is a powerful refrigerated and heating circulator with strong heating and cooling power with an operating temperature range of −25 to +200 °C.
  • Industry-standard LED/TFT display for maximum visibility
  • Adjustable safety circuit
  • Visible and audible alarm
  • Safety drain valve
  • USB/RS232 interfaces (labworldsoft® which enables online updates of device software)
The units proven pressure and suction pump makes the circulator ideal for external applications. The high-quality insulation allows fast heat-up times and reduces heat input at low temperatures. All parameters are manageable (read out, monitored, and completely documented) by software.
CBC 5 control model has optional external solenoid valves (multi-IO port) and removable WiCo (wireless controller) which enables safe remote operation, e.g. outside of a closed fume hood.
Certifications: Safety classification III (FL, DIN 12876).
Informations de livraison: All devices have a PT100 interface for external temperature control and an integrated cooling coil.