Imprimer
Safety lockout hasp, blank
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
225196 225198 265399
BRDY225196EA 0 EUR
BRDY225196 BRDY225198 BRDY265399
Safety lockout hasp, blank
Cadenas de sécurité
These blank lockout hasps can easily be customised with labels.
  • Made from aluminium
  • Available in several colours
  • Six padlocks can be applied
Max. shackle diameter: 8,00 mm
Min. shackle clearance: 20,00 mm