Imprimer
Emergency stop safety covers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
104600 104601
BRDY104600EA 112 EUR
BRDY104600 BRDY104601
Emergency stop safety covers
Cadenas de sécurité
These steel devices guard against unauthorised operation or accidental activation.
  • Can be permanently installed over push/pull or twist-release emergency stops
  • Durable metal bar locks button in the depressed (OFF) position when lock shackle is inserted
  • For use with standard IEC 22,50 mm or NEMA 30,5 mm buttons