Imprimer
Electrician lockout kits
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
196229 196230 196219 196220 196217 196227
BRDY196229EA 223 EUR
BRDY196229 BRDY196230 BRDY196219 BRDY196220 BRDY196217 BRDY196227
Electrician lockout kits
Cadenas de sécurité
These portable kits contain different components.
Electrician lockout kit - contents:
1×Miniature circuit breaker lockout pin-on-standard (PIS)
1×Miniature circuit breaker lockout pin-out-standard (POS)
1× Miniature circuit breaker lockout pin-out-wide (POW)
1×Miniature circuit breaker lockout tie bar (TBLO)
1×No hole circuit breaker 277 V
1×No hole circuit breaker 480 to 600 V
1×Cable lockout device
1×Nylon lockout hasp
1×Large lockout pouch
1×Pen
Electrician lockout kit + padlocks - contents:
2×Full nylon red safety padlocks
1×Miniature circuit breaker lockout pin-on-standard (PIS)
1×Miniature circuit breaker lockout pin-out-standard (POS)
1×Miniature circuit breaker lockout pin-out-wide (POW)
1×Miniature circuit breaker lockout tie bar (TBLO)
1×No hole circuit breaker 277 V
1×No hole circuit breaker 480 to 600 V
1×Cable lockout device
1×Nylon lockout hasp
1×Large lockout pouch
1×Pen
Informations de livraison: The French and Dutch kits additionally include mini tags in French or Dutch.