Imprimer
Ground protection accessories
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28340 28352 28356 28358 28362
119-0316EA 62.2 EUR
119-0316 119-0317 119-0318 119-0319 119-0320
Ground protection accessories
Matériaux absorbants
These ground protection accessories improve berm longevity without sacrificing functionality.
  • Ground mats create a smooth, uniform deployment area with this heavy-duty woven fabric
  • Track mats position inside the berm for extra protection along tire paths
  • Track runners place this high visibility, yellow vinyl coated fabric inside the berm for moderate protection against tire abrasion