Imprimer
Drum storage, EcoPolyBlend™ DrumSheds™
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28675 28676
119-0387EA 5800 EUR
119-0387 119-0388
Drum storage, EcoPolyBlend™ DrumSheds™
Stockage des fûts
A robust design made of 55% recycled polyethylene features convenient rolltop doors to allow access from front or rear.
  • Eco-friendly, weather-resistant drum storage
  • Spacious interior for funnels or pumps
  • Forklift pockets front and back for easy transport
  • Ships fully assembled
Weather-resistant outdoor sheds offer high performance spill control. Forklift pockets on the front and back enable easy transport and relocations. The leakproof pallet sump features thick, durable deck grating that can be removed for easy inspection and clean up of spills. WideEn savoir plus