Imprimer
Personal padlock kit
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
873871 873872 874074
BRDY873871EA 19.1 EUR
BRDY873871 BRDY873872 BRDY874074
Personal padlock kit
Cadenas de sécurité
This padlock kit includes the equipment needed to implement lockout safety procedures. A convenient way to carry padlocks, hasps and keys in a handy pouch.
  • Pouch has a clear face allowing for easy identification
  • Heavy duty retractable key holder features a Kevlar® cord with split ring attachment and a convertible belt clip/belt loop back attachment
  • Key holder is extendable to 1200 mm
  • Clip can be attached to pouch or be used independently
Informations de livraison: The kit includes: One Safety lockout hasp (25 mm shackle width), one Heavy Duty retractable key holder and two red safety padlocks.