Imprimer
Boom diverters, water-filled
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28450 28452 28454 28456
119-0344EA 97 EUR
119-0344 119-0345 119-0346 119-0347
Boom diverters, water-filled
Matériaux absorbants
These water-filled boom diverters temporarily divert and control fluids for easy clean-up or recovery processes. The flexible PVC film construction forms a diversionary barrier also on uneven surfaces such as asphalt or concrete to form a diversionary barrier away from storm drains or as a holding area for wet vacuum operations.
  • Easy-open push plug for filling with standard water hose
  • Redirects liquids away from work areas and storm drains
Water filled booms are used in storm water management, portable washdown/pressure washing diversion, spill diversion, and equipment and personnel decontamination.
Certifications: Helps to comply with EPA for stormwater regulations.