Imprimer
Maintenance spill berms
Fournisseur: JUSTRITE MANUFACTURING
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28400 28402 28404 28406 28408 28410 28412 28414 28416 28418 28421 28422 28424 28426
119-0326EA 239 EUR
119-0326 119-0327 119-0328 119-0329 119-0330 119-0331 119-0332 119-0333 119-0334 119-0335 119-0336 119-0337 119-0338 119-0339
Maintenance spill berms
Matériaux absorbants
Compact and flexible spill berms quickly contain minor spills and leaks during temporary storage of drums, machine parts, batteries, and equipment. Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner.
  • No assembly required; simply unfold and use
  • Encased urethane foam sidewalls offer easy roll-through access without the need for a ramp
  • Walls quickly spring back into place with no memory loss
Lightweight EnGuard™I, PVC coated fabric provides broad UV and chemical resistance. Anchor berm in place using the brass grommets in each corner. Anchor stakes not included. Order optional grates to keep products and machinery above fluid.
Certifications: Helps to comply with EPA SPCC regulations
Informations de commande: Anchor stakes not included. Order optional grates to keep products and machinery above fluid.