Imprimer
Recombinant rLysozyme™
Marques : Merck Millipore (Novagen)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
71110-3 71110-4 71110-5
71110-3EA 84.1 EUR
71110-3 71110-4 71110-5
Recombinant rLysozyme™
Enzymes
rLysozyme™ solution contains a highly purified and stabilised recombinant lysozyme that can be used for lysis of gram negative bacteria, such as E. coli.