Imprimer
Containers, stainless steel
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A416-1 A416-10 A416-20 A416-5 A416-8 A416-2 A416-15
300-0408EA 685 EUR
300-0408 300-0409 300-0412 300-0413 300-0414 300-0411 300-0410
Containers, stainless steel
Récipients de collecte d'échantillon Conteneurs à usage universel
These containers are made of 316L stainless. The high quality crevice-free finish makes these containers ideal for powders, liquids and most demanding applications.
  • Multipurpose containers
  • Robust construction
  • All welds ground and polished
  • Standard 102 or 51 mm lid (Tri-ferrule style)
  • Complete with lid, silicone gasket and clamp band
The handy 101,6 mm tri clamp connection on the top of the containers can be used to clamp on to existing similar equipment.