Imprimer
Lens cleaning tissues
Fournisseur: Ahlstrom-Munksjö
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
146099 146100 146102 146103 146104
BINZ146099EA 2.8 EUR
BINZ146099 BINZ146100 BINZ146102 BINZ146103 BINZ146104
Lens cleaning tissues
Lingettes Lingettes et essuie-mains à usage unique Lingettes optiques
These very fine lens cleaning tissues are made of pure manila vegetable fibre.
  • Non abrasive
Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.
12 g/m²