Imprimer
Volumetric flask, class A, glass, with stopper
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3.668.00214 3.668.00314 3.668.005 3.668.050 3.668.060 3.668.150.BL
WITG3.668.00214EA 14.6 EUR
WITG3.668.00214 WITG3.668.00314 WITG3.668.005 WITG3.668.050 WITG3.668.060 WITG3.668.150.BL
Volumetric flask, class A, glass, with stopper
Ballons Fioles jaugées
Borosilicate glass 3.3, with hollow glass stopper.
  • Very good chemical resistance
  • Very good temperature resistance
Certifications: DIN EN ISO 1042 and DE-M marked.